Learn More
Eppendorf™ CryoCube F570h ULT Freezer, 570 L, VIP, AC, DoLe, 5, (CH), LN2 Backup System
GREENER_CHOICE
Eppendorf CryoCube F570 ULT freezers offer a combination of minimal operating costs and superior sample protection. With a five-compartment layout and five inner doors for easy sample access, the solid polyurethane foam shields your valuable samples.
Brand:  Eppendorf™ F571343037
16889.99 CHF valid until 2025-12-31
Use promo code "25041" to get your promotional price.
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 17358613
  • 17996.70CHF / Each
Only null left
Estimated Shipment: 25-09-2025
Add to basket
Description
Description
  • Volume: 570 L (20.1 ft³), e.g. up to 40,000 samples
  • Efficient insulation, composed of traditional insulation (n-version) and vacuum insulation panels (h-version), saves energy
  • Efficient compressor control system reduces cycle times to lower energy consumption and increases freezer longevity
  • Automatic vent port on front door of upright CryoCube models improves energy consumption and enables fast sample access
  • Lockable main switch to prevent switch-offs by accident
  • Location for chart recorder for data documentation
  • Ergonomic door handle of CryoCube models based on Eppendorf PhysioCare
  • Concept™ requires less force and provides easier access
  • Air-cooled as well as water-cooled versions for further energy saving
  • Stainless steel interior for easy cleaning during maintenance and defrost cycles; powder-coated steel exterior
TRUSTED_SUSTAINABILITY
Specifications
Specifications
570 L
Right
5
Yes
89.8 x 110.3 x 195.1 cm (35.4 x 43.4 x 76.8 in.)
No
CH
7400 W
R290 / R170
5
Vacuum
Classic Interface
2
57.5 x 76.5 x 126.5 cm (22.6 x 30.1 x 49.8 in.)
7 A
230 V
2-stage
25
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Promotions
Promotions