Learn More
CPLX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifica
Antigene CPLX2
Concentrazione 0.33 mg/mL
Applicazioni Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classificazione Polyclonal
Coniugato Unconjugated
Visualizza altre specifiche
Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
16818204
Vedere i documenti
Proteintech
18149-1-AP-20UL
20 μL
145.00€
20µL
16808204
Vedere i documenti
Proteintech
18149-1-AP-150UL
150 μL
410.00€
150µL
Descrizione
Descrizione
Proteins encoded by the complexin/synaphin gene family are cytosolic proteins that function in synaptic vesicle exocytosis. These proteins bind syntaxin, part of the SNAP receptor. The protein product of this gene binds to the SNAP receptor complex and disrupts it, allowing transmitter release. Two transcript variants encoding the same protein have been found for this gene.
Specifica
Specifica
CPLX2
Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Mouse, Human, Rat
P84086, P84087, Q6PUV4
10814, 116657, 12890
CPLX2 Fusion Protein Ag12838
Primary
-20°C
CPLX2
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
921 L, complexin 2, Complexin II, CPLX2, CPX 2, CPX II, CPX2, Hfb1, Synaphin 1
CPLX2
IgG
Antigen Affinity Chromatography