missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPS8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA557682
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: VGLPVEEAVK GILEQGWQAD STTRMVLPRK PVAGALDVSF NKFIPLSEPA P Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
COPS8 is a COP9 signalosome (CSN), which is an evolutionarily conserved protein complex of the eight subunits that interacts with deubiquitinating enzymes and protein kinases and is highly homologous to the lid sub-complex of 26S proteasome. The CSN complex is an essential regulator of the ubiquitin conjugation pathway by mediating the deneddylation of the SCF-type E3 ligase complexes, which leads to a decrease in ubiquitin ligase activity of SCF-complexes such as SCF, CSA or DDB2. It is also involved in phosphorylation of p53, c-jun/JUN, ITPK1 and IRF8/ICSBP, possibly via its association with CK2 and PKD kinases. CSN8 encodes the smallest and the least conserved but first identified subunit of CSN. Recent studies show CSN8 is essential for Drosophila development and is essential for peripheral T cell homeostasis and antigen receptor-induced entry into the cell cycle from quiescence.
Specifica
COPS8 | |
Polyclonal | |
Unconjugated | |
Cops8 | |
9430009J09Rik; AA408242; COP9; COP9 (constitutive photomorphogenic) homolog, subunit 8; COP9 (constitutive photomorphogenic) homolog, subunit 8 (Arabidopsis thaliana); COP9 constitutive photomorphogenic homolog subunit 8; COP9 homolog; COP9 signalosome complex subunit 8; COP9 signalosome subunit 8; COPS8; Csn8; hCOP9; JAB1-containing signalosome subunit 8; SGN8; signalosome subunit 8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10920 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99627 | |
Cops8 | |
Recombinant protein corresponding to Human COPS8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto