Learn More
Fisherbrand™ Cooling of Drain Water for Machines without High Speed Module and Condenser Recycle
The drain water is mixed with cold water before entering the drain.
Descrizione
- Water is cooled to ≤ 60°C
Specifica
Cooling Drain Water |