Learn More
3M™ Protective Coverall 4532+, Cool White
Coverall has a unique specialist material treatment which offers enhanced liquid hold out with added oil and alcohol repellency. Certified to Type 5 for hazardous particles and Type 6 for limited chemical splash, 4532+ coverall provides soft breathable comfort of an SSMMS with the improved liquid performance normally associated with a laminate.
Marca: 3M™ 7100016018
133.92 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Descrizione
- Unique liquid repellency with oil and alcohol resistance material treatment
- Soft and strong multi-layered 52gsm SSMMS for strong performance
- Made from super breathable SSMMS non-woven for effective release of body heat and moisture and reduced heat stress
- 3 panel hood, reinforced triangular gusset panel, 2-way zip and storm flap with sealable tape, elasticated back and ankles to better freedom of movement
- Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
- EN 1073-2 nuclear particulate hold out
- Knitted cuffs and elastic waist and ankles for a better fit and freedom of movement

Specifica
Protective Coveralls | |
Unisex | |
White | |
Elastic | |
Front Zipper | |
CE Category III , EN 1073-2 radioactive particles , EN 1149-5 - electrostatic properties , EN 13034: 2005 + A1: 2009 Type 6 liquid splash , EN ISO 13982-1: 2004 + A1: 2010 Type 5 harmful particles |
4X-Large | |
SSMMS | |
Single-use | |
Knit | |
Non-sterile | |
CE Category III, EN 1073-2, EN 1149-5, EN 13034:2005+A1:2009 Type 6, EN ISO 13982-1:2004 Type 5 |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.