Learn More
Testo™ ComSoft CFR 21 Part 11 Software
Use this software for the programming and readout of Testo data loggers. Testo™ ComSoft CFR 21 Part 11 Software is designed specifically for the requirements of the pharmaceutical industry.
Marca:  Testo™ 0554 1705
Codice prodotto. 11869100
  • 2720.00€ / Pezzo
Solo null disponibili
Spedizione stimata: 13-08-2025
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Validatable software
  • Fulfills all requirements of the FDA (Food and Drug Administration) in a closed system
  • Conformity with the CFR guidelines is confirmed by an independent institute
  • Supports Testo data loggers from the instrument series testo 174, testo 175, testo 176 and testo 184
  • Runs on all 32 and 64-Bit operating systems Windows XP SP2, Windows Vista, Windows 7 (except the respective HOME Edition). Further operating systems available upon request
  • Administration of user groups by administrator
  • Storage of raw data in tamper-proof file format
  • Recognition of carry-over errors thanks to test sums
  • Inactivity lockout to prevent unauthorized access
  • Monitoring of login/logout activity, successful/unsuccessful use of digital signatures and alteration of raw data with the help of an Audit Trail
Specifica
Specifica
Software, ComSoft CFR 21 Part 11
Pharmaceutical applications
Data logger series testo 174, testo 175, testo 176 and testo 184