Learn More
Xylem Analytics™ WTW™ Type-800 Combined Ion-Selective Electrodes (ISEs)
WTW™ Type-800 Combined Ion-Selective Electrodes (ISEs) for measurement of the electrochemical concentration of selected ions. Easy to handle, compact, combined electrodes allow the measurement of small sample volumes. A cost-effective option for analyzing small amounts of material.
Marca: Xylem Analytics™ 106666
1288.00 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Descrizione
- Product Type: Type-800 Combined Electrodes
- Use: Measurement of Electrochemical Ion Concentrations
- Connector: DIN or BNC
- Cable Length: 1 m (3.3 ft.)
Specifica
BNC | |
1 m (3.3 ft.) | |
F 800 BNC | |
Combination ISE |
Requirements: ELY/BR/503 reference electrolyte, ionic strength adapter TISAB, standard solution ES/F | |
5 to 7 | |
F-, Al, PO¾-, Li+ Combined ISE | |
0.02 mg/L-saturation |