Descrizione
- One product = two uses
- Upper protection
- Panoramic vision
- Non-slip bridge
- Comfortable, non-slip straight temples
- Temple or strap version with interchangeable foam
- Pivoting temples
Specifica
Specifica
Certificazione/Conformità | Frame: EN 166 BT CE; Lens marking: 5-1.4 1 BT CE; Chemical risk - Liquid droplets - 3 (Goggle, adapted spectacles with strap or face shield mandatory); Mechanical risk - Low energy impact - F; Mechanical risk- High speed particles at extreme temperatures |
Lens Coating | Anti-fog, Scratch-resistant |
Descrizione | COBRA wrap around safety glasses |
Da utilizzare con (apparecchiatura) | Black |
Include | B-ESP filters; Scratch-resistant, anti-fog (ASAF) |
N. per confezione | 1 ciasc. |
Peso (sistema metrico) | 25 g |
Lens Material | Panoramic vision |
Materiale del telaio | Wraparound, non-slip bridge |
Videos
-
Bolle Safety Cobra
Documenti
Product Information
Specification Sheets
COBRA
Dimensione file: 1.2 MB
Titolo del prodotto
Bollé Safety Cobra ESP Safety Glasses >