Learn More
Zwiebel™ Class F1 Weight Set from 1 to 500 mg
Zwiebel™ Weight Set is intended for use in the verification or calibration of class F2 weights.
Marca: Zwiebel™ PBI56830
628.20 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Includes the following weight: 1 x 1 mg, 2 x 2 mg, 1 x 5 mg, 1 x 10 mg, 2 x 20 mg, 1 x 50 mg, 1 x 100 mg, 2 x 200 mg, 1 x 500 mg
- Triangle wire weights are 1, 10, and 100 mg
- Square and double square wire weights are 2, 20, and 200 mg
- Pentagon wire weights are 5, 50, and 500 mg
- Weights intended for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy class I and high accuracy class II
Specifica
Weight Set | |
1 to 500 mg | |
12 Weights |
OIML R111 Recommendation | |
8500 ±100 kg/m3 |