Learn More
Witeg™ CF-10 High-Performance Centrifuge
Capable of speeds up to 13500 rpm with rotors included.
Brand: Witeg™ DH.WCF00010
This item is not returnable.
View return policy
Description
- Ideal for plasmid DNA and RNA work, protein work, cell culture, cell harvesting, sub-cellular separations and many other applications
- Direct driven brushless DC motor, quiet operation
- Air cooling system
- Digital feedback speed control
- Useful exclusive spin-down function
- Rotor lid for running and reducing noise
- Extremely compact design with small footprint
- Acceleration and braking times lower than 15 seconds
- Storage function for the set values RPM and time
- Body made of ABS, rotor made of PE, lid of PC
- Backlit LCD display
- CE/GMP certified and unique serial number for tracing
Specifications
13,500 rpm | |
109 mm | |
57 dB | |
6 x 0.2/0.5 mL tube adapter | |
CF-10 | |
Fixed Angle |
Backlit LCD | |
178 mm | |
150 mm | |
Digital feedback control | |
Yes |