Learn More
JP Selecta™ Centromix II-BL Centrifuge
Used to obtain autologous extract of blood tissue, adipose and bone marrow.
Brand: JP Selecta™ 7002555
1361.60 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Used in the sanitary, cosmetic and plastic surgery fields
- Important help for serious injuries treatment and for damages tissues regeneration
- Induction drive motor
- External DUR-AL case, with AISI 304 stainless steel bowl
Specifications
23 cm | |
Centrifuge | |
35 cm |
30 cm | |
120mL max volume | |
180 w |