Learn More
Erlab™ Captair™ Smart 834 Ductless Fume Hood
Designed with advanced carbon filtration to minimize chemical exposure.
Specifica
Tipo di filtro | BE+ |
---|---|
Da utilizzare con (applicazione) | For liquid products and powders |
Descrizione
- Sensors to detect filter breakthrough of solvents, acids or formaldehyde
- Safety back up filter in case of main filter saturation
- Continuous monitoring of air face velocity
Erlab Safety Program
- Application analysis and validation, usage certification, filter change reminders
- Connect with eGuard™ for real-time status, notifications and alerts
- Smart-Light feature for lighting up the hood
Flexibility
- Configurable filtration column
- No ductwork needed
Environment
- No dangerous chemicals are released into atmosphere
- Low energy consumption
Specifica
BE+ | |
For liquid products and powders |