Learn More
Color Coded pH Buffers in Twin Neck Bottle Format, Reagecon™
Sfoglia Tutto Reagecon Prodotti
Click to view available options
Quantità:
250 mL
500 ml
1 L
pH:
10 at 20°C
10 at 25°C
7 at 20°C
7 at 25°C
Descrizione
- Colored pH Buffer Solutions
- Tested at 20°C or 25°C
- Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
- NIST traceable
- Presented in various pack sizes
- No possibility of contamination
- No need for separate measuring container for use in the calibration of the Electrode
- Correct quantity of buffer required for calibration is dispensed into the calibrating chamber giving rise to no waste
- Ideally suited for field work
- Easy to carry
- 250ml, 500ml and 1L sizes available
Specifica
Specifica
Colore | Blue |
pH | 10 at 20°C |
Quantità | 1 L |
Imballaggio | HDPE Twin Neck Bottle |