Learn More
Alpaco™ Fascette serratubo in ottone
Provides uniform tightening around tubing for a watertight seal. Alpaco™ Brass Clamps with Vertical Screws are guaranteed waterproof and hand-lock without any tools.
Marca:  Alpaco™ M-NH 17/10-1159
Codice prodotto. 11904947
  • 28.18€ / pezzi 10
Solo null disponibili
In stock
Edge
Aggiungi al carrello
Descrizione
Descrizione
  • Reusable
  • Resists pressure
  • No lubricant required for assembly
  • Smooth clamping to avoid tubing damage
Specifica
Specifica
Confezione da 10
Brass
12 to 17 mm
Clip
Hose O.D.: 12 to 17 mm