Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Shaft, interchangeable
Use as an alternative to magnetic stirrers with high viscosity or high stirring speeds.
Marca:  DWK Life Sciences 286307205
Codice prodotto. 15804648
  • 1220.00€ / pezzi 50
Solo null disponibili
Spedizione stimata: 12-08-2025
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Solid DURAN™ KPG™ stirrer shafts are used for further processing into a wide variety of stirrers. They are available in the qualities ground and polished. The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
  • Solid
  • Available in polished and ground version
  • The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding
TRUSTED_SUSTAINABILITY
Specifica
Specifica
50/Pk.
Stirring of reaction solutions with high viscosities or at high stirring speeds; For further processing to various stirrer shapes and types
Polished