Learn More
Bel-Art™ SP Scienceware™ Miniature Vial Caddy
Provides compact, organized storage and transport.
Brand: Bel-Art™ H18911-0150
This item is not returnable.
View return policy
Description
- For 5 to 8mL miniature vials
- Holds miniature vials up to 17mm dia.
- 150 places (166 pins)
- Side cut-outs for easy grip and carry
- Heat and chemical resistant
- HDPE
- Autoclavable at 121°C (250°F)
Specifications
Side cut-outs | |
Autoclavable | |
HDPE | |
Holding 5 to 8mL miniature vials | |
32.4 x 21 x 4.8cm (12.75 x 8.75 x 1.875 in.) |
Miniature Vial Caddy | |
Vials | |
150 (166 Pins) | |
1/Pk. |