Learn More
behr Labor-Technik™ behrotest™ Sampling Scoop
The behrotest™sampling scoop with telescopic rod and beaker glass is suitable for sampling accessible water flows with sufficient depth.
Brand:  behr Labor-Technik™ B00196083
Product Code. 10471981
  • 735.00 € / Each
Estimated Shipment: 27-05-2025
Explore more special offers
Add to basket
Description
Description
  • Stable, three-piece holding rod with telescopic insertions
    • Length is infinitely adjustable between 1 m and 3 m
      • Infinitely adjustable clamping device
        • Beaker holder made of stainless steel
Specifications
Specifications
Stainless Steel
Sampling scoop, 3-piece, length 375 cm, stainless steel flask, 1 L
3-piece
375 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions