Learn More
Base Oil 75, SPEX CertiPrep™
Sfoglia Tutto SPEX Certiprep Prodotti
Click to view available options
Quantità:
500 mL
1 gal.
Dimensione della confezione:
3.80L
500mL
Descrizione
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Base Oil 75 Blank
- Certified Base Oil
- Organometallic Oil Standard
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifica
Specifica
Materiale o nome chimico | Base Oil 75 Blank |
Imballaggio | Triple-leached LDPE bottle |
Certificato di analisi | With Certificate of Analysis |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
Da utilizzare con (apparecchiatura) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Tipo | Certified Reference Material |
Solvent or Matrix | Base Oil 75 |
Industry Type | Petrochemical Testing |
Quantità | 500 mL |
Product intended for laboratory use only