Learn More
Axygen™ Axypet™ 8-Channel Digital Pipettor, Calibration 3x4 ALT
High levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.
710.00€
Specifica
Certificazione/Conformità | ISO17025 |
---|---|
Numero di canali | 8 |
Product Line | Axypet |
Tipo | Multi-channel Pipettor |
Quantità | 1/Pk., 1/Cs |
Codice del prodotto | Marca | Volume (sistema metrico) | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Volume (sistema metrico) | Prezzo | Quantità e disponibilità | |||||
17213634
|
Axygen™
AP-8-10-ALT |
0.5 to 10 μL | N/A | ||||||
17273634
|
Axygen™
AP-8-50-ALT |
5 to 50 μL | N/A | ||||||
17233634
|
Axygen™
AP-8-200-ALT |
Da 20 a 200 μl | N/A | ||||||
17253634
|
Axygen™
AP-8-300-ALT |
50 to 300 μL | N/A | ||||||
Descrizione
- ISO 17025 Pre-calibrated
- Thumb-friendly soft spring pipetting system reduces fatigue
- Unique side EZIject button option reduces thumb movement
- Finger-hook provides for a more relaxed grip
- Large ergonomic push button is easy on the thumb during contact
- Locking feature prevents accidental volume changes during pipetting procedures
- Push button and ejector button are ergonomically located
- Curved ejector bar allows tips to be ejected in steps, reducing the mount of force required for ejection
- Tip manifold rotates 360°
- Each channel of the pipettor has an individual, precision piston assembly to ensure accuracy and reproducibility from one pipetting series to the next
- Shafts withdraw when pressed against a row of tips
- Each shaft moves independently, minimizing jamming of tips
- Micrometer is continuously adjustable for selection of whole or fractional volumes
Specifica
ISO17025 | |
Axypet | |
1/Pk., 1/Cs |
8 | |
Multi-channel Pipettor |