Learn More
CEEC Controlled Environment Equipment Corporation™ Antibacteria Carpets
Use CEEC Control Environ Eqpt Corp™ Antibacteria Carpets to bond directly to the floor or in aluminum. After use, detach the sheets, which are made of polyethylene.
Marca: CEEC Controlled Environment Equipment Corporation™ 177406
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Eliminate bacterial contamination
- Sheets numbered 30 to 1
- Supplied in cartons of 4 stacks of 30 sheets polyethylene
Specifica
91 x 114 cm | |
120/Pk. | |
Blue |