Learn More
JP Selecta™ Cencom II Angle Rotor Centrifuge
Providing digital electronic control of speed and time.
Brand: JP Selecta™ 7002240
349.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For general laboratory use and teaching
- External case made from coated ABS plastic
- Rubber suspended motor reduces any motor vibrations
- Fixed angle 6-place rotor with identification code
- Transparent polycarbonate lid that allows the user to see inside the centrifuge
- Speed selection from 1000 to 4000rpm in 500rpm steps
- Time selection from 1 to 60min in steps of 1
- Push button start and stop
- Supplied complete with 6 x 15mL tubes with adapters and 6 adapters for 15mL Falcon tubes
- Safety micro-switch that disconnects the power to the motor when the lid is opened
Specifications
6 x 15 mL | |
1,760 x g | |
6 x 15mL tubes with adapters | |
Yes | |
Speed and time |
4,000 rpm | |
28 cm | |
80 w | |
4.5 kg | |
Fixed Angle |