Learn More
Aluminum oxide, single crystal, Thermo Scientific™
Codice prodotto. 15441647
Click to view available options
Quantità:
1 Pc.
5 Pc.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 1344-28-1
Formula molecolare Al2O3
Molecular Weight (g/mol) 101.96
Numero MDL MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Sinonimo aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminum;oxygen(2-)
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
Specifica
Materiale o nome chimico Aluminum oxide
Quantità 5 Pc.
Imballaggio Shrink-wrapped