Learn More
Fisherbrand™ Aluminum Scaffolding Tubes
Provides for sturdy scaffolding construction in the laboratory. Fisherbrand™ Aluminum Scaffolding Tubes are manufactured from a durable aluminum alloy.
Marca: Fisherbrand™ 2121/1
Description
- Approx. 13mm diameter
- Available in two different lengths
- Fits with Fisherbrand scaffolding connectors and foot adapters
Specifications
Aluminum Alloy | |
1 m | |
Tube |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Aluminum Scaffolding Tubes > Length: 1m
Spot an opportunity for improvement?Share a Content Correction