Learn More
JP Selecta™ Stainless Steel Dissecting Curved Forceps
Suitable for general laboratory purpose. JP Selecta™ Stainless Steel Dissecting Curved Forceps is made of AISI 304 stainless steel.
8.00€
Specifications
Product Type | Dissecting Forceps |
---|---|
Material | Stainless Steel (AISI 304) |
Shape | Curved |
Length (Metric) | 140 mm |
Product Code | Brand | Tip Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tip Style | Price | Quantity & Availability | |||||
12042834
|
JP Selecta™
1000865 |
Round |
8.00€
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
|||||
12032834
|
JP Selecta™
1000864 |
Fine |
8.00€
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
|||||
Description
- Made of AISI 304 stainless steel
Specifications
Dissecting Forceps | |
Stainless Steel (AISI 304) | |
Curved | |
140 mm |