Learn More
Krüss Optronic™ Abbe AR4 Analogue Refractometer
Brand: Krüss Optronic™ AR4
1034.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Specifications
Analog Refractometer | |
±0.0002nD | |
110/220 V |
1.3 to 1.7 | |
ASTM D1218 | |
110/220 V |