Learn More
AADACL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14021-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Neutral cholesterol ester hydrolase 1 (NCEH) also known as arylacetamide deacetylase-like 1 (AADACL1) or KIAA1363 is an enzyme that in humans is encoded by the NCEH1 gene. NCEH is an enzyme located in the endoplasmic reticulum. NCEH hydrolyzes 2-acetyl monoalkylglycerol ether, as part of an enzymatic pathway regulating the levels of platelet activating factor and lysophospholipids that may be involved in cancer development.Specifica
AADACL1 | |
Polyclonal | |
Unconjugated | |
NCEH1 | |
AADACL1, KIAA1363, KIAA1363, NCEH1,NCEH, NCEH, NCEH1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
294930, 320024, 57552 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B2GV54, Q6PIU2, Q8BLF1 | |
Nceh1 | |
AADACL1 Fusion Protein Ag5136 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |