Learn More
Tocris Bioscience™ A 83-01
GREENER_CHOICE
Codice prodotto. 18025492
Click to view available options
Quantità:
10 mg
50 mg
Dimensione della confezione:
10mg
50mg
Selective inhibitor of TGF-βRI, ALK4 and ALK7
A 83-01 is a potent inhibitor of TGF-β type I receptor ALK5 kinase, type I activin/nodal receptor ALK4 and type I nodal receptor ALK7 (IC50 values are 12, 45 and 7.5 nM respectively). A 83-01 blocks phosphorylation of Smad2 and inhibits TGF-β-induced epithelial-to-mesenchymal transition. Only weakly inhibits ALK-1, -2, -3, -6 and MAPK activity. More potent than SB 431542 (Cat. No. 1614). A 83-01 inhibits differentiation of rat induced pluripotent stem cells (riPSCs) and increases clonal expansion efficiency. Helps maintain homogeneity and long-term in vitro self-renewal of human iPSCs. Also promotes neural differentiation of hPSCs as part of a chemical cocktail.
For more information about how A 83-01 may be used, see our protocol: Converting Fibroblasts into Cardiomyocytes (9C Cocktail)
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 909910-43-6
Formula molecolare C25H19N5S
Peso molecolare 421.522
Specifica
Materiale o nome chimico A 83-01
Quantità 50 mg
Peso formulazione 421.52
Purezza >98%