Learn More
7-Fluoroindole, 97%
Codice prodotto. p-7047108
Click to view available options
Quantità:
250 mg
1 g
Dimensione della confezione:
1g
250mg
CAS: 387-44-0 | C8H6FN | 135.14 g/mol
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in methanol. (almost transparency)
Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 387-44-0
Formula molecolare C8H6FN
Molecular Weight (g/mol) 135.14
Numero MDL MFCD01074502
InChI Key XONKJZDHGCMRRF-UHFFFAOYSA-N
Sinonimo 7-fluoroindole, 1h-indole, 7-fluoro, pubchem7294, 7-fluoranyl-1h-indole, acmc-209j1q, ksc222e4r, 7-fluoro-1h-indole, buttpark 24\07-35
PubChem CID 2774504
IUPAC Name 7-fluoro-1H-indole
SMILES FC1=C2NC=CC2=CC=C1
Specifica
Materiale o nome chimico 7-Fluoroindole
Punto di fusione 61°C to 63°C
Quantità 250 mg
Beilstein 114740
Informazioni di solubilità Soluble in methanol. (almost transparency)
Peso formulazione 135.14
Percent Purity 97%