Learn More
5-Acetylsalicylic acid, 98+%
Codice prodotto. p-7081617
Click to view available options
Quantità:
5 g
25 g
100 g
Dimensione della confezione:
100g
25g
5g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
CAS: 13110-96-8 | C9H8O4 | 180.159 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 13110-96-8
Formula molecolare C9H8O4
Molecular Weight (g/mol) 180.159
Numero MDL MFCD00013978
InChI Key NZRDKNBIPVLNHA-UHFFFAOYSA-N
PubChem CID 83151
IUPAC Name 5-acetyl-2-hydroxybenzoic acid
SMILES CC(=O)C1=CC(=C(C=C1)O)C(=O)O
Specifica
Materiale o nome chimico 5-Acetylsalicylic acid
Punto di fusione 214°C to 216°C
Punto d'infiammabilità 250°C (482°F)
Quantità 5 g
Beilstein 2096968
Sensibilità Moisture sensitive
Sinonimo 5-acetylsalicylic acid, 3-acetyl-6-hydroxybenzoic acid, 5-acetyl-2-hydroxy-benzoic acid, benzoic acid, 5-acetyl-2-hydroxy, 5-acetyl salicylic acid, acmc-1boxk, 5-acetylsalicylicacid, 5-acetyl-2-hydroxybenzoic acid, benzoic acid,5-acetyl-2-hydroxy
Peso formulazione 180.15
Percent Purity ≥98%