Learn More
4-n-Pentylcyclohexanone, 98+%
Codice prodotto. 11459587
Click to view available options
Quantità:
10 g
50 g
Dimensione della confezione:
10g
50g
CAS: 61203-83-6 | C11H20O | 168.28 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 61203-83-6
Formula molecolare C11H20O
Molecular Weight (g/mol) 168.28
Numero MDL MFCD00059581
InChI Key UKLNPJDLSPMJMQ-UHFFFAOYSA-N
Sinonimo 4-pentylcyclohexanone, 4-n-pentylcyclohexanone, 4-amylcyclohexanone, 4-n-amylcyclohexanone, 4-pentyl-1-cyclohexanone, 4-pentylcyclo hexanone, pubchem20247, acmc-209mq3, uklnpjdlspmjmq-uhfffaoysa
PubChem CID 566097
IUPAC Name 4-pentylcyclohexan-1-one
SMILES CCCCCC1CCC(=O)CC1
Specifica
Materiale o nome chimico 4-n-Pentylcyclohexanone
Densità 0.89
Quantità 10 g
Numero UN UN3082
Peso formulazione 168.28
Percent Purity ≥98%