Learn More
Sigma Laborzentrifugen™ 4-5L Centrifuge
This centrifuge has been designed specifically targeting the requirement for high-throughput in clinical applications.
Brand: Sigma Laborzentrifugen™ 1040590
This item is not returnable.
View return policy
Description
- New motor with high torque, fast acceleration
- Optimized air circulation design for efficient rotor cooling, low noise
- Complete set includes rotor, buckets, sealing lids and adapters for 9-15mL tubes
- Speed range up to 4700rpm
- Optimized for swing-out applications
- Robust components
- Pneumatic spring support lid
- Maximum capacity of 4 x 650mL
- Maintenance-free induction drive motor
- Up to 20g imbalance tolerance
- Suitable for low speeds, e.g. for cell sedimentation, accuracy ±1rpm
- Window in the centrifuge lid for external speed control
Specifications
67 dB | |
496 mm | |
634 mm |
Max. acc. 46 seconds; max. dec. 26 seconds | |
390 mm | |
1010 w |