Learn More
3-Methylbenzeneboronic acid, 97%
Codice prodotto. 11466206
Click to view available options
Quantità:
1 g
5 g
25 g
Dimensione della confezione:
1g
25g
5g
CAS: 17933-03-8 | C7H9BO2 | 135.96 g/mol
Suzuki coupling reactions
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 17933-03-8
Formula molecolare C7H9BO2
Molecular Weight (g/mol) 135.96
Numero MDL MFCD00040198
InChI Key BJQCPCFFYBKRLM-UHFFFAOYSA-N
Sinonimo 3-tolylboronic acid, m-tolylboronic acid, 3-methylphenyl boronic acid, 3-methylbenzeneboronic acid, 3-methyl phenyl boronic acid, 3-tolyboronic acid, 3-methylphenyl boranediol, m-methylphenylboronic acid, m-tolyl boronic acid
PubChem CID 2733950
IUPAC Name (3-methylphenyl)boronic acid
SMILES CC1=CC=CC(=C1)B(O)O
Specifica
Materiale o nome chimico 3-Methylbenzeneboronic acid
Punto di fusione ∼160°C to 162°C
Quantità 5 g
Beilstein 2935968
Peso formulazione 135.96
Percent Purity 97%