Learn More
3-Dimethylaminobenzoyl chloride hydrochloride, tech. 90%
Codice prodotto. p-7047268
Click to view available options
Quantità:
1 g
5 g
Dimensione della confezione:
1g
5g
CAS: 117500-61-5 | C9H11Cl2NO | 220.093 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 117500-61-5
Formula molecolare C9H11Cl2NO
Molecular Weight (g/mol) 220.093
Numero MDL MFCD02094023
InChI Key NUDQPATXNFWFNK-UHFFFAOYSA-N
Sinonimo 3-dimethylaminobenzoyl chloride hydrochloride, 3-dimethylamino benzoyl chloride hydrochloride, acmc-20an50, 3-dimethylaminobenzoyl chloride hcl, 3-dimethylamino-benzoyl chloride hydrochloride salt, 3-dimethylamino benzoyl chloride-hydrogen chloride 1/1
PubChem CID 22732309
IUPAC Name 3-(dimethylamino)benzoyl chloride;hydrochloride
SMILES CN(C)C1=CC=CC(=C1)C(=O)Cl.Cl
Specifica
Materiale o nome chimico 3-Dimethylaminobenzoyl chloride hydrochloride
Punto di fusione 131°C to 133°C
Quantità 1 g
Numero UN UN3261
Beilstein 2803337
Sensibilità Moisture sensitive
Peso formulazione 220.1
Percent Purity 90%
Grado Caratteristiche