Learn More
2000 mOsmol/kg calibration standard, Gonotec™
Click to view available options
For Use With: Osmomat 030 Freezing point osmometer
Specifications
Specifications
Concentration | 2000 mOsmol/kg |
For Use With (Equipment) | Osmomat 030 Freezing Point Osmometer |