Learn More
2-(Trifluoromethyl)cyclohexanone, 97%
Codice prodotto. 15446237
Click to view available options
Quantità:
250 mg
1 g
Dimensione della confezione:
1g
250mg
CAS: 56734-74-8 | C7H9F3O | 166.14 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 56734-74-8
Formula molecolare C7H9F3O
Molecular Weight (g/mol) 166.14
Numero MDL MFCD00102125
InChI Key RZSQHJQIBOWYSQ-UHFFFAOYNA-N
Sinonimo 2-trifluoromethyl cyclohexanone, 2-trifluoromethylcyclohexanone, 2-trifluoromethyl cyclohexan-1-one, trifluoromethylcyclohexanone, 2-trifluoromethyl-cyclohexan-1-one
PubChem CID 2777488
IUPAC Name 2-(trifluoromethyl)cyclohexan-1-one
SMILES FC(F)(F)C1CCCCC1=O
Specifica
Materiale o nome chimico 2-(Trifluoromethyl)cyclohexanone
Punti di ebollizione 71°C to 73°C (12 mmHg)
Quantità 1 g
Numero UN UN1224
Peso formulazione 166.14
Percent Purity 97%