Learn More
2-Furoylacetonitrile, 97%
Codice prodotto. p-7021951
Click to view available options
Quantità:
1 g
5 g
25 g
Dimensione della confezione:
1g
25g
5g
CAS: 31909-58-7 | C7H5NO2 | 135.12 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 31909-58-7
Formula molecolare C7H5NO2
Molecular Weight (g/mol) 135.12
Numero MDL MFCD00052210
InChI Key RZNSHBXVTAHWPP-UHFFFAOYSA-N
Sinonimo 2-furoylacetonitrile, 3-furan-2-yl-3-oxopropanenitrile, 3-2-furyl-3-oxopropanenitrile, 2-cyanoacetylfuran, 2-furanoylacetonitrile, 2-furoyl acetonitrile, pubchem11959, acmc-1cock, 2-2-furanoyl acetonitrile
PubChem CID 141671
IUPAC Name 3-(furan-2-yl)-3-oxopropanenitrile
SMILES O=C(CC#N)C1=CC=CO1
Specifica
Materiale o nome chimico 2-Furoylacetonitrile
Punto di fusione 77°C to 80°C
Quantità 1 g
Beilstein 111902
Peso formulazione 135.12
Percent Purity 97%