Learn More
2,6-Difluoro-4-iodophenol, 99%
Codice prodotto. p-7031117
Click to view available options
Quantità:
1 g
10 g
Dimensione della confezione:
10g
1g
CAS: 950858-06-7 | C6H3F2IO | 255.99 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 950858-06-7
Formula molecolare C6H3F2IO
Molecular Weight (g/mol) 255.99
Numero MDL MFCD09907951
InChI Key LHUADYKKTXZZMQ-UHFFFAOYSA-N
PubChem CID 45790937
IUPAC Name 2,6-difluoro-4-iodophenol
SMILES C1=C(C=C(C(=C1F)O)F)I
Specifica
Materiale o nome chimico 2,6-Difluoro-4-iodophenol
Punto di fusione 70°C to 73°C
Quantità 1 g
Sensibilità Light sensitive
Informazioni di solubilità Insoluble in water.
Peso formulazione 255.99
Percent Purity 99%